Anti-TLX2 Rabbit Polyclonal Antibody
Catalog # AVIVARP39235_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:T-cell leukemia homeobox 2
- Antigen symbol:TLX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3196
- Antigen synonyms:HOX11L1|NCX
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TLX2 (T-cell leukemia homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: