Anti-EZH2 Rabbit Polyclonal Antibody
Catalog # AVIVARP38470_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:enhancer of zeste 2 polycomb repressive complex 2 subunit
- Antigen symbol:EZH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2146
- Antigen synonyms:KMT6|EZH2b|WVS|ENX1|ENX-1|KMT6A|WVS2|EZH1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EZH2 (enhancer of zeste homolog 2 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: