Anti-RHOXF2 Rabbit Polyclonal Antibody
Catalog # AVIVARP37789_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Rhox Homeobox Family, member 2
- Antigen symbol:RHOXF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:84528
- Antigen synonyms:CT107|PEPP-2|THG1|PEPP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RHOXF2 (Rhox homeobox family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human