Anti-KCNK13 Rabbit Polyclonal Antibody
Catalog # AVIVARP35347_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:potassium channel, two pore domain subfamily K, member 13
- Antigen symbol:KCNK13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:56659
- Antigen synonyms:THIK-1|THIK1|K2p13.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KCNK13 (potassium channel, subfamily K, member 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Pig