Anti-KCNIP4 Rabbit Polyclonal Antibody
Catalog # AVIVARP34621_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Kv channel interacting protein 4
- Antigen symbol:KCNIP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:80333
- Antigen synonyms:CALP|KCHIP4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KCNIP4 (Kv channel interacting protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: