Anti-DMTF1 Rabbit Polyclonal Antibody
Catalog # AVIVARP34603_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:cyclin D binding myb-like transcription factor 1
- Antigen symbol:DMTF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:9988
- Antigen synonyms:MRUL|DMP1|DMTF|hDMP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: DMTF1 (cyclin D binding myb-like transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: