Anti-CORO1A Rabbit Polyclonal Antibody
Catalog # BOSSBS-8550R
Anbieter: Bioss
Technische Daten
- Antibody type:Primary
- Antigen name:Coronin 1A
- Antigen symbol:CORO1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:HCORO1|IMD8|p57|CLABP|CLIPINA|TACO
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: CORO1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: