Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-REG1A Rabbit Polyclonal Antibody
  BOSSBS-6183R
 :  Bioss
Anti-REG1A Rabbit Polyclonal Antibody
  BOSSBS-6183R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Regenerating islet-derived 1 alpha
  • Antigen symbol:
    REG1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Antigen synonyms:
    PSP|P19|PTP|ICRF|PSPS|PSPS1|REG
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: REG1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human