Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-ACR Rabbit Polyclonal Antibody
  BOSSBS-5151R
 :  Bioss
undefined
Anti-ACR Rabbit Polyclonal Antibody
  BOSSBS-5151R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Acrosin
  • Antigen symbol:
    ACR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: ACR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat