Anti-GC Rabbit Polyclonal Antibody
Catalog # BOSSBS-3915R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Group-specific component
- Antigen symbol:GC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:VDBP|GRD3|DBP/GC|DBP|VDBG|HEL-S-51
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Vitamin D Binding protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat