Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD44 Rabbit Polyclonal Antibody
Anti-CD44 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2507R
Supplier:  Bioss
undefined
Anti-CD44 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2507R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD44 molecule (Indian blood group)
  • Antigen symbol:
    CD44
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Antigen synonyms:
    MDU2|HUTCH-I|LHR|MC56|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: CD44
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: