Anti-TNFRSF9 Rabbit Polyclonal Antibody
Codice catalogo BOSSBS-2449R
Nome del fornitore: Bioss
Specifiche
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 9
- Antigen symbol:TNFRSF9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:AA408498|Ly63|CD137|A930040I11Rik|ILA|CDw137|AI325004|4-1BB
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: TNFRSF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat