Anti-TNFSF13B Rabbit Polyclonal Antibody
Catalog # BOSSBS-2431R
Anbieter: Bioss
Technische Daten
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Superfamily Member 13B
- Antigen symbol:TNFSF13B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:CD257|ZTNF4|THANK|TALL-1|B-cell Activating Factor|TNFSF20|DTL|TALL1|TNLG7A|BLYS|BAFF
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: TNFSF13B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat