Anti-KIR2DL1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2419R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Killer cell immunoglobulin-like Receptor two Domains long cytoplasmic tail 1
- Antigen symbol:KIR2DL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:NKAT-1|p58.1|NKAT1|CD158A|KIR-K64|NKAT|KIR221
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: KIR2DL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human