Anti-STAT3 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1658R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Signal Transducer And Activator of Transcription 3
- Antigen symbol:STAT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:Phospho-Signal Transducers and Activators of Transcription 3|pY705|AW109958|signal transducer and activator of transcription 3|ADMIO|Aprf|HIES|APRF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: STAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig