Anti-HPSE Rabbit Polyclonal Antibody
Catalog # BOSSBS-1541R
Supplier: Bioss
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:heparanase
- Antigen-Symbol:HPSE
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Pig,Human,Dog,Rat,Mouse
- Western blot:Yes
- Kreuzadsorption:none
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µl
- VE:100 µl
Specifications
About this item
Type: Primary
Antigen: HPSE (heparanase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Pig, Rat