Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-GM-CSFR alpha Rabbit polyclonal antibody
Catalog # BOSSBS-1457R
Supplier:  Bioss
undefined
Anti-GM-CSFR alpha Rabbit polyclonal antibody
Catalog # BOSSBS-1457R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GM-CSFR alpha
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: GM-CSFR alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat