Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-BRG-1/SMARCA4/SNF2 beta Rabbit polyclonal antibody
Catalog # BOSSBS-1408R
Anbieter:  Bioss
Anti-BRG-1/SMARCA4/SNF2 beta Rabbit polyclonal antibody
Catalog # BOSSBS-1408R
Anbieter:  Bioss

Technische Daten

  • Antibody type:
    Primary
  • Antigen symbol:
    BRG1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Antigen synonyms:
    SMARCA4|SNF2 beta
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Technische Daten

Über diesen Artikel

Type: Primary
Antigen: BRG-1/SMARCA4/SNF2 beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat