Anti-MT1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1328R
Anbieter: Bioss
Technische Daten
- Antibody type:Primary
- Antigen name:Melatonin Receptor 1A
- Antigen symbol:MTNR1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:MT-I|Mt-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: MT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat