Anti-Integrin Alpha 5 + Beta 3 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1310R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 5 + Integrin Beta 3
- Antigen symbol:ITGA5 + ITGB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Integrin Alpha 5 + Beta 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat