Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-CUX2 Rabbit Polyclonal Antibody
  BOSSBS-11832R
 :  Bioss
undefined
Anti-CUX2 Rabbit Polyclonal Antibody
  BOSSBS-11832R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Cut-like homeobox 2
  • Antigen symbol:
    CUX2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Antigen synonyms:
    CUTL2|CDP2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: CUX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: