Order Entry
Switzerland
ContactUsLinkComponent
Anti-Elastin Rabbit Polyclonal Antibody
Catalog # BOSSBS-11057R
Anbieter:  Bioss
Anti-Elastin Rabbit Polyclonal Antibody
Catalog # BOSSBS-11057R
Anbieter:  Bioss

Technische Daten

  • Antibody type:
    Primary
  • Antigen name:
    Elastin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Technische Daten

Über diesen Artikel

Type: Primary
Antigen: Elastin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat