Anti-DAB2 Rabbit Polyclonal Antibody
BOSSBS-11044R
: Bioss
- Antikörper-Typ:Primär
- Antigen-Name:Dab, mitogen-responsive phosphoprotein, homolog 2 (Drosophila)
- Antigen-Symbol:DAB2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Kreuzadsorption:none
- Antigen-Synonyme:DOC-2|differentially-expressed protein 2|FLJ26626|DOC2|disabled homolog 2|mitogen-responsive phosphoprotein
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- VE:100 µl
Type: Primary
Antigen: DAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat