Anti-ABCC2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1092R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding cassette, Subfamily C (CFTR/MRP), Member 2
- Antigen symbol:ABCC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:DJS|cMRP|MRP2|CMOAT|ABC30
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ABCC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat