Anti-ADRB3 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1063R
Supplier: Bioss
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Adrenoceptor Beta 3
- Antigen-Symbol:ADRB3
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Kreuzadsorption:none
- Antigen-Synonyme:Adrb-3|FLJ99960|BETA3AR
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- VE:100 µl
Specifications
About this item
Type: Primary
Antigen: ADRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat