Anti-CASP9 Rabbit Polyclonal Antibody
Catalog # BOSSBS-0049R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Caspase 9
- Antigen symbol:CASP9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Dog