Specifications
- Antibody type:Primary
- Antigen name:Synuclein beta
- Antigen symbol:SNCB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Gene ID:6620
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli (HIS-beta-Synuclein)
- Purification:Affinity purification
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
Synuclein is a small, soluble protein primarily expressed in neural tissue. The synuclein family includes three known proteins: α-synuclein, β-synuclein, and γ-synuclein.
α-Synuclein is a very conserved, small acidic protein (140 residues) with a molecular weight of approximately 19 kDa that accounts for about 1% of total protein in neurons. It is concentrated primarily in presynaptic axon terminals and plays a central role in initiating Parkinson’s disease. It is the principal constituent of Lewy bodies which are formed by protein aggregation and inclusion body formation.
α-Synuclein lacks secondary or tertiary structure, so it belongs to the family of natively unfolded proteins, many of which act as chaperones. Much of this α-synuclein has been post-translationally modified to increase the rate of oligomerization.
Mutant forms of α-Synuclein are associated with autosomal dominant Parkinson’s disease. The mutations increase the aggregation rate of the resultant α-synuclein
β-Synuclein is found primarily in brain tissue and is seen mainly in presynaptic terminals. It is predominantly expressed in the neocortex, hippocampus, striatum, thalamus, and cerebellum.
β-Synuclein may protect the central nervous system from the neurotoxic effects of α-synuclein by inhibiting aggregation of α-synuclein which occurs in neurodegenerative diseases such as Parkinson's disease.
γ-Synuclein is a synuclein protein found primarily in the peripheral nervous system (in primary sensory neurons, sympathetic neurons, and motor neurons) and retina. γ-Synuclein expression in breast tumors is a marker for tumor progression.
Western Blot: 1:2,000;
Type: Primary
Antigen: Synuclein (synuclein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat