Anti-mRNA capping enzyme Mouse Monoclonal Antibody [clone: T16-AF3F1]
Catalog # ABFRLF-MA0353
Supplier: AbFrontier
Specifications
- Antibody type:Primary
- Antigen name:RNA guanylyltransferase and 5'-phosphatase
- Antigen symbol:RNGTT
- Clonality:Monoclonal
- Clone:T16-AF3F1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Gene ID:8732
- Antigen synonyms:HCE|HCE1|CAP1A|hCAP
- Storage temperature:Store for 1 year at –20 °C from date of shipmen
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli
- Purification:Ammonium sulfate precipitation
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
Western Blot: 1:2,000;
Type: Primary
Antigen: mRNA capping enzyme (mRNA capping enzyme)
Clonality: Monoclonal
Clone: T16-AF3F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human