Technische Daten
- Antibody type:Primary
- Antigen name:NCK adaptor protein 2
- Antigen symbol:NCK2
- Clonality:Monoclonal
- Clone:AF3F11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Gene ID:8440
- Antigen synonyms:GRB4|NCKbeta
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli (His-NCK2)
- Purification:Protein G purified
- Size:100 µl
- Pk:0,1 mL
Technische Daten
Über diesen Artikel
NCK1 is one of the adaptor proteins which mediate specific protein-protein interactions in signaling processes. Adaptor proteins usually contain several domains like SH2 (Src homology 2) and SH3 which allow specific interactions with other specific proteins.
NCK1 and NCK2 showing high sequence identity (68%) have three SH3 domains and a C-terminal SH2 domain. Both of them bind receptor tyrosine kinases such as PDGFR and other tyrosine phosphorylated
proteins via their SH2 domains. Various molecules which interact with SH domains of Nck and regulate signaling process of actin cytoskeleton reorganization have been identified. Ncks are thought to have important functions in the development of mesodermal structures during embryogenesis, linked to a role in cell movement and cytoskeletal reorganization.
Nck also have a function in modulating mRNA translation at the level of initiation by interacting eukayotic initiation factor 2 (eIF2). Under the stressed conditions, protein synthesis is reduced by inhibiting the activity of eIF2 through the phosphorylation, transiently inhibiting recycling of eIF2
into its active form.
Western Blot: 0.5 ?g/ml; Immunoprecipitation: 2.0 ?g;
Type: Primary
Antigen: NCK2 (NCK adaptor protein 2)
Clonality: Monoclonal
Clone: AF3F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat