Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Tumor protein p53
- Antigen-Symbol:P53
- Klonalität:Monoklonal
- Klon:AF2C1
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Immunpräzipitation:Yes
- Isotyp:IgG1
- Reaktivität:Human
- Western blot:Yes
- Kreuzadsorption:none
- Gen-ID:7157
- Antigen-Synonyme:P53|BCC7|TRP53|LFS1
- Lagertemperatur:Store for 1 year at –20 °C from date of shipment
- Konzentration:1 mg/ml
- Transporttemperatur:Ice
- Immunogen:Recombinant human protein purified from E.coli (His-p53)
- Reinigung:Protein G purified
- Größe:100 µl
- VE:0,1 mL
Specifications
About this item
p53 is a transcription factor that regulates the cell cycle and hence functions as a tumor suppressor. p53 has been described as "the guardian of the genome", referring to its role in conserving stability by preventing genome mutation. p53 has many anti-cancer mechanisms: activating DNA repair proteins when DNA has sustained damage, holding the cell cycle at the G1/S regulation point on DNA damage recognition, initiating apoptosis if the DNA damage proves to be irrepairable. Human p53 is 393 amino acids long and has three domains: N-terminal transcription-activation domain (TAD), which activates transcription factors. 2) central DNA-binding core domain (DBD) 3) C-terminal homo-oligomerisation domain (OD); tetramerization greatly increases the activity of p53 in vivo. Mutations that deactivate p53 in cancer usually occur in the DBD and most of these mutations destroy the ability of the protein to bind to its target DNA sequences.
Western Blot: 0.5 ?g/ml; Immunoprecipitation: 2.0 ?g;
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: Monoclonal
Clone: AF2C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human