Order Entry
Switzerland
ContactUsLinkComponent
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 48D2]
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 48D2]
Catalog # ABFRLF-MA0175
Supplier:  AbFrontier
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 48D2]
Catalog # ABFRLF-MA0175
Supplier:  AbFrontier

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
  • Antigen symbol:
    SERPINA1
  • Clonality:
    Monoclonal
  • Clone:
    48D2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    5265
  • Antigen synonyms:
    alpha1AT|PRO2275|AAT|PI|A1A|PI1|A1AT
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein purified from Human plasma
  • Purification:
    Protein G purified
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

α1-antitrypsin, 52kDa, is the prototypic member of the serine protease inhibitor (serpin) superfamily, which have a major role in inactivating neutrophil elastase and other proteases to maintain protease-antiprotease balance. The protein is synthesized mainly in the liver, but exerts its main function in the lung where it inhibits a variety of serine proteases, but primarily neutrophil elastase. It provides more than 90% of the antiproteolytic shield of the lower respiratory tract protecting the elastic tissue from degradation. Disorders of the enzyme include α 1-antitrypsin deficiency, a hereditary disorder in which lack of α 1-antitrypsin leads to uninhibited tissue breakdown during inflammation. This causes pulmonary emphysema and leads to liver cirrhosis in severe cases.

Western Blot: 0.1 ?g/ml; Immunohistochemistry: Paraffin-embedded sections;

Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: Monoclonal
Clone: 48D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human