Specifications
- Antibody type:Primary
- Antigen name:Glycosylphosphatidylinositol specific phospholipase D1
- Antigen symbol:GPLD1
- Clonality:Monoclonal
- Clone:38A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Gene ID:2822
- Antigen synonyms:PLD|PIGPLD|GPIPLDM|GPIPLD|PIGPLD1
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Shipping temperature:Ice
- Immunogen:Protein purified from Human plasma
- Purification:Ammonium sulfate precipitation
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
GPI-PLD (glycosylphosphati- dylinositol-specific phospholipase D), a 815-amino acid protein, is expressed in numerous tissues and cells and specifically cleaves GPI-anchored proteins. Liver has the highest level of GPI-PLD expression and is the primary organ contributing to GPI-PLD in the serum. GPI-PLD is abundant in serum in which it associates with apolipoproteins AI and AIV. Increased serum GPI-PLD is associated with insulin resistance and elevated serum triglycerides. Many surface proteins are attached to eukaryotic cell membranes via glycosylphosphatidylinositol (GPI) anchors that are covalently bound to the C-terminus of the protein and cleavage of the GPI moiety by GPI-PLD, only enzyme known that cleavage GPI anchor, may represent a means of regulating attachment of these proteins to the cell surface, or alternatively, their release into the extracellular environment.
Western Blot: 1:500; Immunohistochemistry: Paraffin-embedded sections;
Type: Primary
Antigen: GPLD1 (glycosylphosphatidylinositol specific phospholipase D1)
Clonality: Monoclonal
Clone: 38A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human