Order Entry
Switzerland
ContactUsLinkComponent
Anti-Fibronectin Mouse Monoclonal Antibody [clone: 16E5]
Anti-Fibronectin Mouse Monoclonal Antibody [clone: 16E5]
Catalog # ABFRLF-MA0151
Supplier:  AbFrontier
Anti-Fibronectin Mouse Monoclonal Antibody [clone: 16E5]
Catalog # ABFRLF-MA0151
Supplier:  AbFrontier

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibronectin
  • Antigen symbol:
    FN1
  • Clonality:
    Monoclonal
  • Clone:
    16E5
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    2335
  • Antigen synonyms:
    ED-B|FNZ|MSF|CIG|GFND2|LETS|GFND|FN|FINC
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein purified from Human plasma
  • Purification:
    Protein G purified
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

Fibronectin is a high molecular weight glycoprotein containing about 5% carbohydrate that binds to receptor proteins that span the cells membrane, called integrins. In addition to integrins, they also bind extracellular matrix components such as collagen, fibrin and heparin. Fibronectin can be found in the blood plasma in its soluble form which is composed of two 250 kDa subunits joined together by disulfide bonds. Plasma fibronectin is made in the liver by hepatocytes. The insoluble form that serves as a linker in the extracellular matrix is a large complex of cross-linked subunits. Fibronectin is involved in many cellular processes, including tissue repair, embryogenesis, blood clotting, and cell migration/ adhesion, so it can be used as a therapeutic agent for wound healing. It is also one of the few proteins for which production increases with age without any associated pathology.

Western Blot: 1.0ug/ml; Immunoprecipitation: 5.0ug;

Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: Monoclonal
Clone: 1600000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human