
Acetic acid ≥80%, EMPROVE® EXPERT for biopharmaceutical production
Catalog # 1.37072.1000
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2790
ADR:8,
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152