Order Entry
Switzerland
ContactUsLinkComponent
Micro Ready-To-Go Cleaning Trolley
Micro Ready-To-Go Cleaning Trolley
Catalog # 141-1216
Supplier:  FHP VILEDA SCS
undefined
Micro Ready-To-Go Cleaning Trolley
Catalog # 141-1216
Supplier:  FHP VILEDA SCS

Specifications

  • Description:
    Micro Ready-to-go cleaning trolley
  • W×D×H:
    670×520×1040 mm
  • Pk:
    1

Specifications

About this item

Small, light and ready-equipped trolley made of lacquered steel.

  • Strong materials and excellent finishing stands every detail of design and quality
  • Solid construction and assembled without tools
  • Adjustable push handles
  • Refuse sack holder
  • Upper holder
  • Mop holder
  • Hooks
  • Wheels 100 mm, one with brake

Micro Ready-to-go is a small and well planned cleaning trolley to suit even the losing urban spaces and narrow aisles. The carriage is adapted to the Origo accessories - buckets and boxes. Optional accessories for Micro Ready-to-go is a bottle holder and a storage shelf for garbage bags. Micro Ready-to-go is tailored to our wide range of professional cleaning equipment.