Autosampler syringes, Dionex™
Catalog # DION053916
Supplier: DIONEX
Specifications
- Volume:250 µl
- Description:Sample syringe, standard
- Pk:1
Specifications
About this item
This syringe is designed to pipette liquid from one vial to another or to waste and mixes a vial by repeatedly drawing and expelling the vial contents. It can also be used to deliver a sample to the injection valve.