Ultrapure water systems, arium® comfort I
It is compact, environmentally friendly, reliable and easy-to-use. arium® comfort I produces ASTM Type 1 ultrapure water and Type 3 pure water. The system contains state of the art reverse osmosis technology and a unique cartridge. arium® comfort I optimizes water consumption using the integrated iJust control unit. Applied for HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC-Analysis, photometry and for buffer solutions.
- Optional integrated TOC monitor
- Compact design
- Flexible display and SD card slot
- Feed water for laboratory devices such as autoclaves and glassware washers
- Optimized, economic water consumption
- Premium product-water quality at all times
- Guaranteed longer life of down-stream ultrapure water systems
The optional integrated UV lamp (185 | 254 nm) avoids microbiological growth and reduces the content of TOC to a minimum. Reproducible and reliable results are guaranteed due to the low TOC content of up to <2 ppb.
- Catalog No:
- VIVAH2O-I-2-TOC-B
- 171-1529
- Model:
- arium® comfort I, with UV lamp and TOC monitor
- arium® comfort I, with UV lamp and TOC monitor
- Flow rate:
- 0,266 l/min
- 0,266 l/min
- TOC content:
- ≤2 ppb
- ≤2 ppb
- Resistivity at +25 °C:
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- Particles:
- <1/ml
- <1/ml
- Microorganisms:
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- W×D×H:
- 435×476×501 mm
- 435×476×501 mm
- Operating weight with full tank:
- ~31 kg
- ~31 kg
- Weight:
- ~23 kg
- ~23 kg
Frequently Bought Together