ContactUsLinkComponent
Qualitative filter papers, fast filtration
Qualitative filter papers, fast filtration
Supplier:  PRAT DUMAS
Certificates
undefined
Qualitative filter papers, fast filtration
Supplier:  PRAT DUMAS
Specifications
  • Catalog No:
  • PRATA009307
  • PRATA015307
  • PRATA007307
  • PRATJ015307
  • Weight:
  • 75 g/m2
  • 75 g/m2
  • 75 g/m2
  • 75 g/m2
  • Thickness:
  • 240 µm
  • 240 µm
  • 240 µm
  • 240 µm
  • Particle retention:
  • 8 - 10 µm
  • 8 - 10 µm
  • 8 - 10 µm
  • 8 - 10 µm