ContactUsLinkComponent
Reusable half masks, DUETTA
  111-3051
undefined
Reusable half masks, DUETTA
  111-3051
  • Type:
    DUETTA with Bayonet lock
  • Size:
    U*
  • Pk:
    1
Reusable half mask with double bayonet filter connection, plastic mask body with soft, sealing surface and elastic adjustment straps with head gear. Protects against dusts, gases and vapours, or their combinations.
  • Excellent field of vision, unrestricted freedom of movement and outstanding comfort
  • Superior balance through double filter technology
  • Optimal sealing provided by newly designed sealing rim
  • Easily dismantled for cleaning and maintenance
  • One-piece headband for optimal fit
  • Non-allergenic
One mask size fits all face shapes
 : EN 140; 89/686/EWG