Carboys, wide neck, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
PP, translucent.
- Good chemical resistance
- Llarge handles for transport and pouring
- Meets the requirements of regulation 177.1520 governing storage of foodstuffs and USP Class VI certified
Ideal for storing and dispensing large batches of culture media, distilled water and other solutions.
Frequently Bought Together