ContactUsLinkComponent
Accessories for micro sieve set Mini-Sieve™, SP Bel-Art
Accessories for micro sieve set Mini-Sieve™, SP Bel-Art
Catalog # 510-9566
undefined
Accessories for micro sieve set Mini-Sieve™, SP Bel-Art
Catalog # 510-9566
Specifications
  • For:
    Micro sieve set, Mini-Sieve™
  • Mesh size:
    25, 35, 45, 60, 80, 120, 170, 230 µm
  • Description:
    Replacement screen inserts, phosphor-bronze
  • Seq:
    02
  • Pk:
    1 SET