ContactUsLinkComponent
Magnetic stirring bar restrainer, Spinbar®, SP Bel-Art
Magnetic stirring bar restrainer, Spinbar®, SP Bel-Art
Catalog # BELAF377861000
undefined
Magnetic stirring bar restrainer, Spinbar®, SP Bel-Art
Catalog # BELAF377861000
Specifications
  • Description:
    Stirring bar restrainer, blue
  • Pk:
    1
Specifications
About this item
The Spinbar® restrainer fits ergonomically in one hand, and when placed alongside the outer wall of a mixing vessel it retains and secures the stirring bar in position.
  • Ensures no direct contact with the stirring magnet
  • Retains magnetic stirring bars up to 80 mm long