ContactUsLinkComponent
VWR® MediaBag TW Dehydrated Culture Media in Bags
VWR® MediaBag TW Dehydrated Culture Media in Bags
Catalog # 7101.00450
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® MediaBag TW Dehydrated Culture Media in Bags
Catalog # 7101.00450
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    100
  • Description:
    MediaBag TW for BPW
  • Sterile water to be added:
    450 ml/bag
  • For:
    450 ml
Specifications
About this item
MediaBags TW (Twirl-bag) are filled with pre-weighed granulated media, double-bagged bag and irradiated. Add sterile water under aseptic conditions.