Order Entry
Switzerland
ContactUsLinkComponent
Petri dish rack, SP Bel-Art
Petri dish rack, SP Bel-Art
Catalog # BELAF189850100
undefined
Petri dish rack, SP Bel-Art
Catalog # BELAF189850100

Specifications

  • Description:
    Petri dish rack, white, holds 54 dishes (100 mm)
  • Pk:
    1

Specifications

About this item

Clear TPX body, PP post. This Petri dish rack holds up to 54 (Ø 100 mm) dishes.

  • Finger slots for easy removal of dishes
  • Built-in handle for easy transportation
  • Rubber feet prevent rack from sliding on the benchtop

W×L×H: 200×340×230 mm.