0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Block Heaters
Digital Block Heaters
Peqlab, Accessories for block heaters, digital, HX-1 and HX-2
Peqlab, Accessories for block heaters, digital, HX-1 and HX-2
Peqlab, Accessories for block heaters, digital, HX-1 and HX-2
Catalog # PEQL91-D102B
Supplier:
VWR Collection
Expand image
Peqlab, Accessories for block heaters, digital, HX-1 and HX-2
Catalog # PEQL91-D102B
Supplier:
VWR Collection
Specifications
Description:
Aluminium block, 48×0,2 ml tubes with pop stopper
Seq:
01
Pk:
1
For:
HX-1/HX-2
Specifications
About this item
Product Details
Product Details