Scoops, Open PharmaScoop®
Supplier: Sampling Systems
Certificates
About this item
These general purpose scoops made from stainless steel 316 feature a heavy duty, crevice free construction which makes them ideal for use in the food and pharmaceutical industries.
- Open top design, flat bottom with rounded edges
- 316 stainless steel is a high-quality grade and is therefore very corrosion resistant
- Satin finish
Frequently Bought Together