Temperature indicating strips
Supplier: Testo
About this item
testoterm measuring strips are self-adhesive films with heat-sensitive elements for checking and monitoring temperature.
- Change in colour in two to three seconds
- Practical booklet format including 10 thermometer strips
- 8 temperature levels
They are used for measurements on moving and/or small parts, for long-term monitoring.
5 Options Available Below
Product Details & Documents
testoterm measuring strips are self-adhesive films with heat-sensitive elements for checking and monitoring temperature.
- Change in colour in two to three seconds
- Practical booklet format including 10 thermometer strips
- 8 temperature levels
They are used for measurements on moving and/or small parts, for long-term monitoring.
Specifications for Product Family
Specifications
- Catalog No:
- TEST0646.0108
- TEST0646.0916
- TEST0646.1724
- TEST0646.2532
- TEST0646.3341
- Storage temperature:
- +25 °C max.; Storage in refrigerator recommended
- +25 °C max.; Storage in refrigerator recommended
- +25 °C max.; Storage in refrigerator recommended
- +25 °C max.; Storage in refrigerator recommended
- +25 °C max.; Storage in refrigerator recommended
- Temperature accuracy:
- +43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value - +43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value - +43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value - +43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value - +43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value
Specifications
Product Family Options
Product Information
- RangeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.TEST0646.0108Storage temperature+25 °C max.; Storage in refrigerator recommendedTemperature accuracy+43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value - Specifications:
More
Specifications
Cat. No.TEST0646.0916Storage temperature+25 °C max.; Storage in refrigerator recommendedTemperature accuracy+43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value - Specifications:
More
Specifications
Cat. No.TEST0646.1724Storage temperature+25 °C max.; Storage in refrigerator recommendedTemperature accuracy+43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value - Specifications:
More
Specifications
Cat. No.TEST0646.2532Storage temperature+25 °C max.; Storage in refrigerator recommendedTemperature accuracy+43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value - Specifications:
More
Specifications
Cat. No.TEST0646.3341Storage temperature+25 °C max.; Storage in refrigerator recommendedTemperature accuracy+43...+154 °C: ±1,5 °C;
from +160 °C: ±1 % ±1 °C of the respective temperature value