Order Entry
Switzerland
ContactUsLinkComponent
Peqlab PerfectBlue Midi ExW, Wide Format Gel System Midi ExW
Peqlab PerfectBlue Midi ExW, Wide Format Gel System Midi ExW
Catalog # 700-0531
Supplier:  VWR Collection
undefined
Peqlab PerfectBlue Midi ExW, Wide Format Gel System Midi ExW
Catalog # 700-0531
Supplier:  VWR Collection

Specifications

  • Description:
    PerfectBlue™ Wide Format Gel System Midi ExW
  • Seq:
    01
  • Model:
    Midi ExW
  • Gel volume:
    160 ml
  • Buffer volume:
    ~1400 ml
  • Run time:
    ≥45 h
  • Gel size W×L:
    230×140 mm
  • Sample capacity:
    25 - 200
  • Unit W×L×H:
    260×290×100 mm
  • Pk:
    1

Specifications

About this item

With its extra wide design, the Midi ExW is ideal when there is a need to run lots of samples (up to 200) just a short distance, for example when genotyping. This format, along with its compact footprint, offers rapid run times without compromising on power. The result is a system for high throughput sample processing generating crisp clear band data. As with all PerfectBlue™ gel systems, gels can be poured directly in the trays at temperatures up to 60 °C. With the Midi ExW, the trays can be walled with effective gasketed 'end gates' which are supplied in the system. Additionally, gel size can be reduced by using wall combs or the aluminium casting dam.

*Based on an agarose gel of approximately 5 mm thickness

Ordering information: Each system is supplied with four microtitre combs (1,5 mm thick, 50 teeth), lid, gel tray, tank, cables and end gates for gel pouring.