Orders LinkContactUsLinkComponent
Rabbit plasma
Supplier:   Biowest
Rabbit plasma
Supplier:  Biowest
Certificates
3 Options Available Below
Product Details & Documents
Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.
Specifications for Product Family
Specifications
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • S2600-100
    Rabbit plasma with EDTA, sterile filtered
    Rabbit plasma with EDTA, sterile filtered
    100 ml
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 36.70
    SFr. 36.70
    1 * 100 mL
       SFr. 36.70
    Specifications:
    More
    Specifications
    Cat. No.
    S2600-100
  • S2600-500
    Rabbit plasma with EDTA, sterile filtered
    Rabbit plasma with EDTA, sterile filtered
    500 ml
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 128.00
    SFr. 128.00
    1 * 500 mL
       SFr. 128.00
    Specifications:
    More
    Specifications
    Cat. No.
    S2600-500
  • S2610-100
    Rabbit plasma with sodium citrate, sterile filtered
    Rabbit plasma with sodium citrate, sterile filtered
    100 ml
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 44.00
    SFr. 44.00
    1 * 100 mL
       SFr. 44.00
    Specifications:
    More
    Specifications
    Cat. No.
    S2610-100