ContactUsLinkComponent
Ammonia solution 20%, NORMATOM®, Ultrapure for trace metal analysis
Corrosive
Irritant
Ammonia solution 20%, NORMATOM®, Ultrapure for trace metal analysis
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Irritant
Ammonia solution 20%, NORMATOM®, Ultrapure for trace metal analysis
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Assay:
    20-22%
  • Ag (Silver):
    Max. 10 ppt
  • Al (Aluminium):
    Max. 20 ppt
  • As (Arsenic):
    Max. 10 ppt
  • Au (Gold):
    Max. 10 ppt
  • Ba (Barium):
    Max. 10 ppt
  • Be (Beryllium):
    Max. 10 ppt
  • Bi (Bismuth):
    Max. 10 ppt
  • Ca (Calcium):
    Max. 10 ppt
  • Cd (Cadmium):
    Max. 10 ppt
  • Ce (Cerium):
    Max. 10 ppt
  • Co (Cobalt):
    Max. 10 ppt
  • Cr (Chromium):
    Max. 10 ppt
  • Cs (Cesium):
    Max. 10 ppt
  • Cu (Copper):
    Max. 10 ppt
  • Dy (Dysprosium):
    Max. 10 ppt
  • Er (Erbium):
    Max. 10 ppt
  • Eu (Europium):
    Max. 10 ppt
  • Fe (Iron):
    Max. 10 ppt
  • Ga (Gallium):
    Max. 10 ppt
  • Gd (Gadolinium):
    Max. 10 ppt
  • Ge (Germanium):
    Max. 10 ppt
  • Hf (Hafnium):
    Information only
  • Hg (Mercury):
    Max. 200 ppt
  • Ho (Holmium):
    Max. 10 ppt
  • In (Indium):
    Max. 10 ppt
  • K (Potassium):
    Max. 10 ppt
  • La (Lanthanum):
    Max. 10 ppt
  • Li (Lithium):
    Max. 10 ppt
  • Lu (Lutetium):
    Max. 10 ppt
  • Mg (Magnesium):
    Max. 10 ppt
  • Mn (Manganese):
    Max. 10 ppt
  • Mo (Molybdenum):
    Max. 10 ppt
  • Na (Sodium):
    Max. 20 ppt
  • Nb (Niobium):
    Max. 10 ppt
  • Nd (Neodymium):
    Max. 10 ppt
  • Ni (Nickel):
    Max. 10 ppt
  • Pb (Lead):
    Max. 10 ppt
  • Pd (Palladium):
    Information only
  • Pr (Praseodymium):
    Max. 10 ppt
  • Pt (Platinum):
    Information only
  • Rb (Rubidium):
    Max. 10 ppt
  • Re (Rhenium):
    Information only
  • Rh (Rhodium):
    Max. 10 ppt
  • Ru (Ruthenium):
    Information only
  • Sb (Antimony):
    Max. 10 ppt
  • Sc (Scandium):
    Max. 10 ppt
  • Se (Selenium):
    Information only
  • Sm (Samarium):
    Max. 10 ppt
  • Sn (Tin):
    Max. 10 ppt
  • Sr (Strontium):
    Max. 10 ppt
  • Tb (Terbium):
    Max. 10 ppt
  • Te (Tellurium):
    Max. 10 ppt
  • Th (Thorium):
    Max. 10 ppt
  • Ti (Titanium):
    Max. 10 ppt
  • Tl (Thallium):
    Max. 10 ppt
  • Tm (Thulium):
    Max. 10 ppt
  • U (Uranium):
    Max. 10 ppt
  • V (Vanadium):
    Max. 10 ppt
  • W (Tungsten):
    Max. 10 ppt
  • Y (Yttrium):
    Max. 10 ppt
  • Yb (Ytterbium):
    Max. 10 ppt
  • Zn (Zinc):
    Max. 10 ppt
  • Zr (Zirconium):
    Max. 10 ppt